missing translation for 'onlineSavingsMsg'
Learn More

Cafestol

Catalog No. p-7043066
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65355MF 50 mg
AAJ65355MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ65355MF Supplier Thermo Scientific Chemicals Supplier No. J65355MF

Natural extract from coffee beans which induces glutathione S-transferase | CAS: 469-83-0 | C20H28O3 | 316.44 g/mol

Natural extract from coffee beans which induces glutathione S-transferase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 469-83-0
Molecular Formula C20H28O3
Molecular Weight (g/mol) 316.44
MDL Number MFCD01075769
InChI Key DNJVYWXIDISQRD-HWUKTEKMSA-N
Synonym cafestol, cafesterol, ccris 1518, unii-ac465t6q6w, 5a,8-methano-5ah-cyclohepta 5,6 naphtho 2,1-b furan-7-methanol, 3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-7-hydroxy-10b-methyl-, 3bs,5as,7r,8r,10ar,10bs, 5a,8-methano-5ah-cyclohepta 5,6 naphtho 2,1-b furan-7-methanol, 3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-7-hydroxy-10b-methyl-, 3bs-3balpha,5abeta,7beta,8beta,10aalpha,10bbeta, --cafestol, 1s,4s,12s,13r,16r,17r-17-hydroxymethyl-12-methyl-8-oxapentacyclo 14.2.1.0 1,13 .0 4,12 .0 5,9 nonadeca-5 9 ,6-dien-17-ol, 3bs,5as,7r,8r,10ar,10bs-7-hydroxymethyl-10b-methyl-3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-5a,8-methanocyclohepta 5,6 naphtho 2,1-b furan-7-ol
PubChem CID 108052
ChEBI CHEBI:3291
IUPAC Name (1S,4S,12S,13R,16R,17R)-17-(hydroxymethyl)-12-methyl-8-oxapentacyclo[14.2.1.0¹,¹³.0⁴,¹².0⁵,⁹]nonadeca-5(9),6-dien-17-ol
SMILES C[C@@]12CCC3=C(C=CO3)[C@H]1CC[C@@]13C[C@@H](CC[C@@H]21)[C@@](O)(CO)C3

Specifications

Melting Point 156°C to 158°C
Color White
Density 1.23
Quantity 50 mg
Merck Index 14,1634
Solubility Information Soluble in dichlormethane, DMSO or ethanol
Formula Weight 316.44
Physical Form Powder
Chemical Name or Material Cafestol
RTECSNumber PB9185090
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only