missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Caerulein, Sulfated

For cellular and molecular biology applications

$427.21 - $427.21

Specifications

Purification Method HPLC
Common Name Ceruletide
Content And Storage -30°C to -10°C
Format Powder
For Use With (Application) Stimulates smooth muscle and increases digestive secretions
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ64320MCR
View Documents
Thermo Scientific Chemicals
J64320MCR
1 mg
Each for $427.21
Only null left
 
Description

Description

Stimulates smooth muscle and increases digestive secretions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

HPLC
-30°C to -10°C
Stimulates smooth muscle and increases digestive secretions
Caerulein, sulfated
Glp-Gln-Asp-Tyr(SO3H)-Thr-Gly-Trp-Met-Asp-Phe-NH2
Ceruletide
Powder
1352.40
RUO - research use only
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only