Learn More
Cadmium tellurite, 99%
CAS: 15851-44-2 | CdO3Te | 288.01 g/mol
$593.84 - $593.84
Chemical Identifiers
| CAS | 15851-44-2 |
|---|---|
| Molecular Formula | CdO3Te |
| Molecular Weight (g/mol) | 288.01 |
| MDL Number | MFCD00049413 |
| InChI Key | GKMPTXZNGKKTDU-UHFFFAOYSA-L |
| Synonym | cadmium tellurite, cadmium 2+ tellurite, cadmium tellurium trioxide, cd.o3te, acmc-20alj5, cadmium 2+ ion tellurite, telluric acid h2teo3 , cadmium salt 1:1, tellurous acid cadmium salt, telluric acid h2teo3 ,cadmium salt 1:1 |
| PubChem CID | 167475 |
| IUPAC Name | cadmium(2+);tellurite |
| SMILES | [Cd++].[O-][Te]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1747614
|
Thermo Scientific Chemicals
A1747614 |
25 g |
Each for $593.84
|
|
|||||
Description
It is employed in the preparation, characterization and structure of Ca8[AlO2]12Te2 and Cd8[AlO2]12Te2; two new members of the sodalite family.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed in the preparation, characterization and structure of Ca8[AlO2]12Te2 and Cd8[AlO2]12Te2; two new members of the sodalite family.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 15851-44-2 | |
| 288.01 | |
| GKMPTXZNGKKTDU-UHFFFAOYSA-L | |
| 167475 | |
| [Cd++].[O-][Te]([O-])=O |
| CdO3Te | |
| MFCD00049413 | |
| cadmium tellurite, cadmium 2+ tellurite, cadmium tellurium trioxide, cd.o3te, acmc-20alj5, cadmium 2+ ion tellurite, telluric acid h2teo3 , cadmium salt 1:1, tellurous acid cadmium salt, telluric acid h2teo3 ,cadmium salt 1:1 | |
| cadmium(2+);tellurite |
Specifications
| 15851-44-2 | |
| CdO3Te | |
| UN2570 | |
| Insoluble in water. | |
| [Cd++].[O-][Te]([O-])=O | |
| 288.01 | |
| 288.01 | |
| Cadmium tellurite |
| 25 g | |
| MFCD00049413 | |
| cadmium tellurite, cadmium 2+ tellurite, cadmium tellurium trioxide, cd.o3te, acmc-20alj5, cadmium 2+ ion tellurite, telluric acid h2teo3 , cadmium salt 1:1, tellurous acid cadmium salt, telluric acid h2teo3 ,cadmium salt 1:1 | |
| GKMPTXZNGKKTDU-UHFFFAOYSA-L | |
| cadmium(2+);tellurite | |
| 167475 | |
| 99% |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber : 239-963-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only