missing translation for 'onlineSavingsMsg'
Learn More

Cadmium sulfide, 99.999% (metals basis)

Catalog No. AA4545530
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4545530 250 g
AA4545518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4545530 Supplier Thermo Scientific Chemicals Supplier No. 04545530

Used as pigment in the chemical industry; photoresistors; optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of this compound are used for the preparation of optical devices operating in the infrared regaion of spectrum. | CAS: 1306-23-6 | CdS | 144.47 g/mol

Cadmium sulfide is used as pigment in the chemical industry; photoresistors; optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of this compound are used for the preparation of optical devices operating in the infrared regaion of spectrum. It is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1306-23-6
Molecular Formula CdS
Molecular Weight (g/mol) 144.47
MDL Number MFCD00010922
InChI Key FRLJSGOEGLARCA-UHFFFAOYSA-N
Synonym cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow
PubChem CID 14783
ChEBI CHEBI:50833
IUPAC Name sulfanylidenecadmium
SMILES [S--].[Cd++]

Specifications

Melting Point 980°C (sublimation)
Physical Form Pieces
Quantity 250 g
UN Number UN3077
Solubility Information Insoluble in water.
Formula Weight 144.46
Percent Purity 99.999%
Packaging Plastic bottle
Specific Gravity 4.82
Density 4.82
Chemical Name or Material Cadmium sulfide
Show More Show Less
Hazard Category H302-H341-H350-H361fd-H372
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 215-147-8
RTECSNumber EV3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only