missing translation for 'onlineSavingsMsg'
Learn More

Cadmium sulfide, 99.99% (metals basis)

Catalog No. AA4151114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4151114 25 g
AA4151122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4151114 Supplier Thermo Scientific Chemicals Supplier No. 04151114

Vacuum deposition | CAS: 1306-23-6 | CdS | 144.47 g/mol

Vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1306-23-6
Molecular Formula CdS
Molecular Weight (g/mol) 144.47
MDL Number MFCD00010922
InChI Key FRLJSGOEGLARCA-UHFFFAOYSA-N
Synonym cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow
PubChem CID 14783
ChEBI CHEBI:50833
IUPAC Name sulfanylidenecadmium
SMILES [S--].[Cd++]

Specifications

Melting Point 980°C (sublimation)
Color Orange
Physical Form Sintered Lump
Quantity 25 g
UN Number UN3077
Merck Index 14,1628
Formula Weight 144.46
Percent Purity 99.99%
Odor Odorless
Density 4.82 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium sulfide
Show More Show Less
Hazard Category H302-H341-H350-H361fd-H372
Hazard Statement GHS H Statement
H350-H372-H341-H361-H302
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 215-147-8
RTECSNumber EV3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only