Learn More
Cadmium sulfide, 98%
CAS: 1306-23-6 | CdS | 144.47 g/mol
Supplier: Thermo Scientific Chemicals A145440E
Description
Cadmium sulfide is used as a pigment in the chemical industry; for manufacturing of photoresistors, optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of cadmium sulfide are used for the preparation of optical devices operating in the infrared regaion of spectrum. Cadmium sulfide is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCadmium sulfide is used as a pigment in the chemical industry; for manufacturing of photoresistors, optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of cadmium sulfide are used for the preparation of optical devices operating in the infrared regaion of spectrum. Cadmium sulfide is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and acids.
Specifications
| Cadmium sulfide | |
| 1306-23-6 | |
| CdS | |
| UN3077 | |
| cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow | |
| FRLJSGOEGLARCA-UHFFFAOYSA-N | |
| sulfanylidenecadmium | |
| 14783 | |
| 144.46 | |
| Odorless |
| 980°C (sublimation) | |
| 2500 g | |
| MFCD00010922 | |
| 14,1628 | |
| Insoluble in water. | |
| [S--].[Cd++] | |
| 144.47 | |
| CHEBI:50833 | |
| 98% | |
| 4.82 |
Chemical Identifiers
| 1306-23-6 | |
| 144.47 | |
| FRLJSGOEGLARCA-UHFFFAOYSA-N | |
| 14783 | |
| [S--].[Cd++] |
| CdS | |
| MFCD00010922 | |
| cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow | |
| CHEBI:50833 |
Safety and Handling
GHS H Statement
H350-H372-H341-H361-H302
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H341-H350-H361fd-H372
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 215-147-8
RTECSNumber : EV3150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only