missing translation for 'onlineSavingsMsg'
Learn More

Cadmium sulfide, 98%

Catalog No. AAA1454436
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1454436 500 g
AAA1454422 100 g
AAA145440E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1454436 Supplier Thermo Scientific Chemicals Supplier No. A1454436
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 1306-23-6 | CdS | 144.47 g/mol

Cadmium sulfide is used as a pigment in the chemical industry; for manufacturing of photoresistors, optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of cadmium sulfide are used for the preparation of optical devices operating in the infrared regaion of spectrum. Cadmium sulfide is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cadmium sulfide is used as a pigment in the chemical industry; for manufacturing of photoresistors, optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of cadmium sulfide are used for the preparation of optical devices operating in the infrared regaion of spectrum. Cadmium sulfide is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and acids.

Chemical Identifiers

CAS 1306-23-6
Molecular Formula CdS
Molecular Weight (g/mol) 144.47
MDL Number MFCD00010922
InChI Key FRLJSGOEGLARCA-UHFFFAOYSA-N
Synonym cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow
PubChem CID 14783
ChEBI CHEBI:50833
SMILES [S--].[Cd++]

Specifications

Melting Point 980°C (sublimation)
Quantity 500 g
UN Number UN3077
Merck Index 14,1628
Solubility Information Insoluble in water.
IUPAC Name cadmium(2+) sulfanediide
Formula Weight 144.46
Percent Purity 98%
Odor Odorless
Density 4.82
Chemical Name or Material Cadmium sulfide
Show More Show Less
Hazard Category H302-H341-H350-H361fd-H372
Hazard Statement GHS H Statement
H350-H372-H341-H361-H302
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 215-147-8
RTECSNumber EV3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only