missing translation for 'onlineSavingsMsg'
Learn More

Cadmium sulfate hydrate, Puratronic™, 99.996% (metals basis)

Catalog No. p-4781830
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1067509 10 g
AA1067518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1067509 Supplier Thermo Scientific Chemicals Supplier No. 01067509
Only null left

CAS: 15244-35-6 | CdO4S | 208.47 g/mol

It is used as an electrolyte in a Weston standard cell, a pigment in fluorescent screens. It is used in the manufacture of cadmium salts of fatty acids, pigments, in electroplating of cadmium in electronic circuits. It act as an intermediate, as an accelerator, in the electrode position of nickel and copper, for stabilizing plastics, in cement formation. It has the ability to absorb hydrochloric acid from waste gases from chemical plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15244-35-6
Molecular Formula CdO4S
Molecular Weight (g/mol) 208.47
MDL Number MFCD00151655
InChI Key QCUOBSQYDGUHHT-UHFFFAOYSA-L
IUPAC Name cadmium(2+) sulfate
SMILES [Cd++].[O-]S([O-])(=O)=O

Specifications

Melting Point >40°C to H2O
Color White
Physical Form Crystals
Quantity 10 g
Assay Percent Range 99.996% (Metals basis)
Solubility Information Soluble in water. Insoluble in alcohol and ethyl acetate.
Formula Weight 208.46 (Anhydrous)
Refractive Index 1.565
Density 3.08 g/cm3
Chemical Name or Material Cadmium sulfate hydrate
Hazard Category H301-H330-H340-H350-H360-H372
Hazard Statement Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 233-331-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only