Learn More
Cadmium sulfate hydrate, Puratronic™, 99.996% (metals basis)
Description
It is used as an electrolyte in a Weston standard cell, a pigment in fluorescent screens. It is used in the manufacture of cadmium salts of fatty acids, pigments, in electroplating of cadmium in electronic circuits. It act as an intermediate, as an accelerator, in the electrode position of nickel and copper, for stabilizing plastics, in cement formation. It has the ability to absorb hydrochloric acid from waste gases from chemical plants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >40°C to H2O |
| Color | White |
| Physical Form | Crystals |
| Quantity | 50 g |
| Assay Percent Range | 99.996% (Metals basis) |
| Solubility Information | Soluble in water. Insoluble in alcohol and ethyl acetate. |
| Formula Weight | 208.46 (Anhydrous) |
| Refractive Index | 1.565 |
| Density | 3.08 |
| Chemical Name or Material | Cadmium sulfate hydrate |
Safety and Handling
| Hazard Category | H301-H330-H340-H350-H360-H372 |
| Hazard Statement | Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c |
| DOTInformation | DOT Class: 6.1, Packaging Group: III |
| EINECSNumber | 233-331-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only