missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Sulfate, 8/3-Hydrate, Crystal, Reagent, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18601184
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601184 45 kg Fiber Drum
18601180 25 g Amber Glass Bottle
18601181 125 g Poly Bottle
18601182 500 g Poly Bottle
18601183 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18601184 Supplier Spectrum Chemical Mfg Cor Supplier No. C105045KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C1050, 7790-84-3, CdSO4·8/3H2O

Spectrum™ Chemical Cadmium Sulfate, 8/3-Hydrate, Crystal, Reagent, ACS , is used widely for the electroplating of cadmium in electronic circuits. It is also a precursor to cadmium-based pigment such as cadmium sulfide. It is also used for electrolyte in a Weston standard cell as well as a pigment in fluorescent screens. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 7790-84-3
Molecular Formula CdH2O5S
Molecular Weight (g/mol) 226.49
InChI Key VRFRVKRTIMIRNG-UHFFFAOYSA-L
IUPAC Name cadmium(2+) hydrate sulfate
SMILES O.[Cd++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 45 kg
UN Number UN2570
Percent Purity 98 to 102%
Grade ACS
Packaging Fiber Drum