missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Sulfate, 8/3-Hydrate, Crystal, Reagent, ACS, 98-102%, Spectrum™ Chemical

C1050, 7790-84-3, CdSO4·8/3H2O

Supplier:  Spectrum Chemical Mfg Cor C1050500GM

 View more versions of this product

Catalog No. 18601182

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Cadmium Sulfate, 8/3-Hydrate, Crystal, Reagent, ACS , is used widely for the electroplating of cadmium in electronic circuits. It is also a precursor to cadmium-based pigment such as cadmium sulfide. It is also used for electrolyte in a Weston standard cell as well as a pigment in fluorescent screens. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Specifications

Specifications

7790-84-3
500 g
UN2570
O.[Cd++].[O-]S([O-])(=O)=O
226.49
ACS
100%
CdH2O5S
VRFRVKRTIMIRNG-UHFFFAOYSA-L
cadmium(2+) hydrate sulfate
98 to 102%
Poly Bottle

Chemical Identifiers

7790-84-3
226.49
cadmium(2+) hydrate sulfate
CdH2O5S
VRFRVKRTIMIRNG-UHFFFAOYSA-L
O.[Cd++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions