missing translation for 'onlineSavingsMsg'
Learn More

Cadmium sulfate, 8/3-hydrate, ACS reagent, crystals

Catalog No. AC403761000
Change view
Click to view available options
Quantity:
100 g
2.5 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC403761000 100 g Glass bottle
AC403760025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC403761000 Supplier Thermo Scientific Chemicals Supplier No. 403761000

CAS: 7790-84-3 | Cd3O12S3·8H2O | 769.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7790-84-3
Molecular Formula Cd3O12S3·8H2O
Molecular Weight (g/mol) 769.51
MDL Number MFCD02253000

Specifications

Melting Point 41.0°C
Color White
Physical Form Crystals
Quantity 100 g
Linear Formula 3CdSO4·8H2O
Merck Index 15,1632
Solubility Information Solubility in water: 1130g/L (20°C). Other solubilities: insoluble in alcohol,acids,acetone and ammonia
Formula Weight 769.51
Percent Purity 98.0 to 102.0%
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Cadmium sulfate, 8/3-hydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
May cause genetic defects.
May damage fertility.
May damage the unborn child.
Fatal if inhaled.
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust

RUO – Research Use Only