missing translation for 'onlineSavingsMsg'
Learn More

Cadmium phosphate (ortho), 99%, Thermo Scientific Chemicals

Catalog No. p-7050985
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4020118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4020118 Supplier Thermo Scientific Chemicals Supplier No. 04020118

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13847-17-1
Molecular Formula Cd3O8P2
Molecular Weight (g/mol) 527.182
MDL Number MFCD00049414
InChI Key NRGIRRZWCDKDMV-UHFFFAOYSA-H
Synonym cadmium phosphate, cadmium orthophosphate, tricadmium diphosphate, cadmium ii phosphate, tricadmium bis phosphate, cadmium phosphte cd3 po4 2, cadmium phosphate ortho, phosphoric acid, cadmium salt 2:3, acmc-20alja, cadmium 2+ diphosphate
PubChem CID 26060
IUPAC Name cadmium(2+);diphosphate
SMILES [O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Cd+2].[Cd+2].[Cd+2]

Specifications

Melting Point ∼1500°C
Physical Form Powder
Quantity 50 g
UN Number UN2570
Solubility Information Insoluble in water
Formula Weight 527.14
Percent Purity 99%
Chemical Name or Material Cadmium phosphate (ortho)
Hazard Category H302+H312+H332-H350
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber 237-581-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only