missing translation for 'onlineSavingsMsg'
Learn More

Cadmium perchlorate hexahydrate

CAS: 10326-28-0 | CdCl2H12O14 | 419.396 g/mol

Supplier:  Thermo Scientific Chemicals 01293630

 View more versions of this product

Catalog No. AA1293630

Only null left
Explore available promotions

Description

Description

It is employed as intermediate in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cadmium perchlorate hexahydrate
10326-28-0
250 g
MFCD00015992
Insoluble in water.
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cd+2]
419.396
419.39 (311.30 Anhydrous)
White
Crystals
CdCl2H12O14
cadmium perchlorate hexahydrate, acmc-1c6hb, cd.2clo4.6h2o, cadmium ii perchlorate hexahydrate
PQJZOTJHMRJOEM-UHFFFAOYSA-L
cadmium(2+);diperchlorate;hexahydrate
71317310
Hygroscopic

Chemical Identifiers

10326-28-0
419.396
PQJZOTJHMRJOEM-UHFFFAOYSA-L
71317310
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cd+2]
CdCl2H12O14
MFCD00015992
cadmium perchlorate hexahydrate, acmc-1c6hb, cd.2clo4.6h2o, cadmium ii perchlorate hexahydrate
cadmium(2+);diperchlorate;hexahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c

H272-H302+H312+H332

DOTInformation : DOT Class: 5.1, Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only