missing translation for 'onlineSavingsMsg'
Learn More

Cadmium perchlorate hexahydrate

Catalog No. aa1293618
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1293618 50 g
AA1293630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1293618 Supplier Thermo Scientific Chemicals Supplier No. 01293618
Only null left

CAS: 10326-28-0 | CdCl2H12O14 | 419.396 g/mol

It is employed as intermediate in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10326-28-0
Molecular Formula CdCl2H12O14
Molecular Weight (g/mol) 419.396
MDL Number MFCD00015992
InChI Key PQJZOTJHMRJOEM-UHFFFAOYSA-L
Synonym cadmium perchlorate hexahydrate, acmc-1c6hb, cd.2clo4.6h2o, cadmium ii perchlorate hexahydrate
PubChem CID 71317310
IUPAC Name cadmium(2+);diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cd+2]

Specifications

Color White
Physical Form Crystals
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 419.39 (311.30 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Cadmium perchlorate hexahydrate
Hazard Category H272-H302+H312+H332
Hazard Statement This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only