Learn More
Cadmium oxide, 99%, pure
CAS: 1306-19-0 | CdO | 128.41 g/mol
$78.91 - $78.91
Chemical Identifiers
| CAS | 1306-19-0 |
|---|---|
| Molecular Formula | CdO |
| Molecular Weight (g/mol) | 128.41 |
| MDL Number | MFCD00010921 |
| InChI Key | CFEAAQFZALKQPA-UHFFFAOYSA-N |
| Synonym | cadmium oxide, cadmium monoxide, cadmium fume, aska-rid, kadmu tlenek polish, caswell no. 136aa, ccris 115, epa pesticide chemical code 236200, dsstox_cid_4715 |
| PubChem CID | 14782 |
| IUPAC Name | oxocadmium |
| SMILES | [O--].[Cd++] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC223792500
|
Thermo Scientific Chemicals
223792500 |
250 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1306-19-0 | |
| 128.41 | |
| CFEAAQFZALKQPA-UHFFFAOYSA-N | |
| 14782 | |
| [O--].[Cd++] |
| CdO | |
| MFCD00010921 | |
| cadmium oxide, cadmium monoxide, cadmium fume, aska-rid, kadmu tlenek polish, caswell no. 136aa, ccris 115, epa pesticide chemical code 236200, dsstox_cid_4715 | |
| oxocadmium |
Specifications
| 1385.0°C | |
| 250 g | |
| CdO | |
| MFCD00010921 | |
| cadmium oxide, cadmium monoxide, cadmium fume, aska-rid, kadmu tlenek polish, caswell no. 136aa, ccris 115, epa pesticide chemical code 236200, dsstox_cid_4715 | |
| CFEAAQFZALKQPA-UHFFFAOYSA-N | |
| oxocadmium | |
| 14782 | |
| 99% | |
| Cadmium oxide |
| 1306-19-0 | |
| 98.5% min. (Complexometry) | |
| CdO | |
| 15,1628 | |
| Solubility in water: insoluble. Other solubilities: soluble in diluted acids and alkalies | |
| [O--].[Cd++] | |
| 128.41 | |
| 128.41 | |
| Pure |
Safety and Handling
GHS H Statement
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
Fatal if inhaled.
May cause cancer.
Suspected
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Danger
EINECSNumber : 215-146-2
RUO – Research Use Only