missing translation for 'onlineSavingsMsg'
Learn More

Cadmium oxide, 99.95% (metals basis)

Catalog No. AA1221936
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1221936 500 g
AA12219A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1221936 Supplier Thermo Scientific Chemicals Supplier No. 01221936
Only null left

CAS: 1306-19-0 | CdO | 128.41 g/mol

Cadmium oxide is used in ceramic glazes, cadmium electroplating baths, pigments, phosphors, electrodes for storage batteries, cadmium salts, and heterogeneous catalysis for dehydrogenation. It is a basic conducting material used to prepare transparent conducting films, which finds use in phototransistors, liquid crystal displays, photodiodes, photovoltaic cells, and antireflection coatings. Electrodeposited cadmium oxide films have excellent electrocatalytic activity towards many organic compounds, ozone and oxygen evolution. CdO is selective in phenol photodegradation. Glassy carbon electrode coated with cadmium oxide film has been used to develop a sensitive and rapid assay method for penicillamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1306-19-0
Molecular Formula CdO
Molecular Weight (g/mol) 128.41
MDL Number MFCD00010921
InChI Key CFEAAQFZALKQPA-UHFFFAOYSA-N
Synonym cadmium oxide, cadmium monoxide, cadmium fume, aska-rid, kadmu tlenek polish, caswell no. 136aa, ccris 115, epa pesticide chemical code 236200, dsstox_cid_4715
PubChem CID 14782
SMILES [O--].[Cd++]

Specifications

Melting Point 1430°C
Color Red to Brown
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.95% (Metals basis)
Solubility Information Soluble in dilute acid. Slowly soluble in ammonium salts. Insoluble in water.
IUPAC Name cadmium(2+) oxidandiide
Formula Weight 128.4
Odor Odorless
Density 8.15 g/cm3
Chemical Name or Material Cadmium oxide
Show More Show Less
Hazard Category H330-H335-H341-H350-H361fd-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P304+P340-P308+P313-P310-P501c
DOTInformation DOT Class: 6.1, Packaging Group: I
EINECSNumber 215-146-2
RTECSNumber EV1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only