missing translation for 'onlineSavingsMsg'
Learn More

Cadmium oxide, 98.9%

Catalog No. AA3323530
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3323530 250 g
AA33235A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3323530 Supplier Thermo Scientific Chemicals Supplier No. 03323530
Only null left

CAS: 1306-19-0 | CdO | 128.41 g/mol

Cadmium oxide is used as an electroplating chemical and in the manufacture of cadmium electrodes. It is a component of silver alloys, phosphors, semiconductors, glass and ceramic glazes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1306-19-0
Molecular Formula CdO
Molecular Weight (g/mol) 128.41
MDL Number MFCD00010921
InChI Key CFEAAQFZALKQPA-UHFFFAOYSA-N
Synonym cadmium oxide, cadmium monoxide, cadmium fume, aska-rid, kadmu tlenek polish, caswell no. 136aa, ccris 115, epa pesticide chemical code 236200, dsstox_cid_4715
PubChem CID 14782
SMILES [O--].[Cd++]

Specifications

Melting Point 1430°C
Color Red to Brown
Physical Form Powder
Quantity 250 g
Assay Percent Range 98.9%
Solubility Information Practically insoluble in water. Soluble in dilute acids,slowly soluble in ammonium salts
IUPAC Name cadmium(2+) oxidandiide
Formula Weight 128.4
Odor Odorless
Density 8.15 g/cm3 at 20°C
Chemical Name or Material Cadmium oxide
Show More Show Less
Hazard Category H330-H335-H341-H350-H361fd-H372
Hazard Statement Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P304+P340-P308+P313-P310-P501c
DOTInformation DOT Class: 6.1, Packaging Group: I
EINECSNumber 215-146-2
RTECSNumber EV1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only