missing translation for 'onlineSavingsMsg'
Learn More

Cadmium nitrate tetrahydrate, Puratronic™, 99.999% (metals basis)

Catalog No. AA1066306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1066306 5 g
AA1066314 25 g
AA1066322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1066306 Supplier Thermo Scientific Chemicals Supplier No. 01066306

CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol

It is used for coloring glass and porcelain, and as a flash powder in photography. It is used as a laboratory reagent, in preparation of cadmium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10022-68-1
Molecular Formula CdH2N2O6
Molecular Weight (g/mol) 238.44
MDL Number MFCD00149626
InChI Key IZIPRBSCGMISKX-UHFFFAOYSA-N
Synonym cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
PubChem CID 56924536
ChEBI CHEBI:86156
SMILES [Cd++].O[N+]([O-])=O.O[N+]([O-])=O

Specifications

Boiling Point 132°C
Melting Point 59°C
Color White
Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Soluble in water,acetone,ethyl acetate,alcohol. Insoluble in nitric acid.
IUPAC Name cadmium(2+);dinitrate;tetrahydrate
Formula Weight 308.47 (236.41 Anhydrous)
Odor Weak
Appearance White crystalline
Density 2.45
Chemical Name or Material Cadmium nitrate tetrahydrate
Show More Show Less
Hazard Category H301-H312-H330-H340-H350-H360-H372
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P302+P352-P304+P340-P308+P313-P310-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 233-710-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only