missing translation for 'onlineSavingsMsg'
Learn More

Cadmium nitrate tetrahydrate, 99.9% (metals basis)

CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol

Supplier:  Thermo Scientific Chemicals 04173230

 View more versions of this product

Catalog No. AA4173230


Only null left
Explore available promotions

Description

Description

Cadmium nitrate tetrahydrate is used in Photographic emulsions, coloring glass and porcelain, laboratory reagent, and the production of other cadmium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

132°C
59°C
White
250 g
(metals basis)
MFCD00149626
14,1622
Soluble in water.(109g/100ml)at 0 C and (326g/100ml)at 60 C.Soluble in ammonia,acetone and alcohol.
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
238.44
CHEBI:86156
99.9%
Cadmium nitrate tetrahydrate
10022-68-1
Crystals
99.9% (Metals basis)
CdH2N2O6
UN3087
cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
IZIPRBSCGMISKX-UHFFFAOYSA-N
cadmium(2+);dinitrate;tetrahydrate
56924536
308.47 (236.41 Anhydrous)
2.45

Chemical Identifiers

10022-68-1
238.44
IZIPRBSCGMISKX-UHFFFAOYSA-N
56924536
cadmium(2+);dinitrate;tetrahydrate
CdH2N2O6
MFCD00149626
cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
CHEBI:86156
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H350-H272-H312-H332

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P302+P352-P304+P340-P308+P313-P310-P312-P330-P363-P501c

H301-H312-H330-H340-H350-H360-H372

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 233-710-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only