missing translation for 'onlineSavingsMsg'
Learn More

Cadmium nitrate tetrahydrate, 99.9% (metals basis)

Catalog No. AA4173218
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4173218 50 g
AA4173230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4173218 Supplier Thermo Scientific Chemicals Supplier No. 04173218

CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol

Cadmium nitrate tetrahydrate is used in Photographic emulsions, coloring glass and porcelain, laboratory reagent, and the production of other cadmium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10022-68-1
Molecular Formula CdH2N2O6
Molecular Weight (g/mol) 238.44
MDL Number MFCD00149626
InChI Key IZIPRBSCGMISKX-UHFFFAOYSA-N
Synonym cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
PubChem CID 56924536
ChEBI CHEBI:86156
IUPAC Name cadmium(2+);dinitrate;tetrahydrate
SMILES [Cd++].O[N+]([O-])=O.O[N+]([O-])=O

Specifications

Boiling Point 132°C
Melting Point 59°C
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 99.9% (Metals basis)
UN Number UN3087
Merck Index 14,1622
Solubility Information Soluble in water.(109g/100ml)at 0 C and (326g/100ml)at 60 C.Soluble in ammonia,acetone and alcohol.
Formula Weight 308.47 (236.41 Anhydrous)
Percent Purity 99.9%
Odor Weak
Density 2.45 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium nitrate tetrahydrate
Show More Show Less
Hazard Category H301-H312-H330-H340-H350-H360-H372
Hazard Statement GHS H Statement
H301-H350-H272-H312-H332
Toxic if swallowed.
May cause cancer.
May intensify fire
oxidizer.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P302+P352-P304+P340-P308+P313-P310-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 233-710-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only