Learn More
Cadmium nitrate tetrahydrate, 98.5% min
CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol
Supplier: Thermo Scientific Chemicals 01222836
Description
Cadmium nitrate is used in manufacturing of cadmium salts, photographic emulsions, laboratory reagent and coloring glass. It is used for porcelain and as a flash powder in photography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 132°C | |
| 59°C | |
| White | |
| 500 g | |
| CdH2N2O6 | |
| cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate | |
| IZIPRBSCGMISKX-UHFFFAOYSA-N | |
| cadmium(2+);dinitrate;tetrahydrate | |
| 56924536 | |
| 308.47 (236.41 Anhydrous) | |
| 2.45 g/cm3 |
| Cadmium nitrate tetrahydrate | |
| 10022-68-1 | |
| Crystals | |
| 98.5% min. | |
| MFCD00149626 | |
| Highly soluble in water. Soluble in alcohol,acetone,and ammonia. | |
| [Cd++].O[N+]([O-])=O.O[N+]([O-])=O | |
| 238.44 | |
| CHEBI:86156 | |
| Weak |
Chemical Identifiers
| 10022-68-1 | |
| 238.44 | |
| IZIPRBSCGMISKX-UHFFFAOYSA-N | |
| 56924536 | |
| cadmium(2+);dinitrate;tetrahydrate |
| CdH2N2O6 | |
| MFCD00149626 | |
| cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate | |
| CHEBI:86156 | |
| [Cd++].O[N+]([O-])=O.O[N+]([O-])=O |
Safety and Handling
GHS H Statement
H301-H350-H272-H312-H332
P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P302+P352-P304+P340-P308+P313-P310-P312-P330-P363-P501c
H301-H312-H330-H340-H350-H360-H372
DOTInformation : DOT Class: 6.1; Packaging Group: II
EINECSNumber : 233-710-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only