missing translation for 'onlineSavingsMsg'
Learn More

Cadmium nitrate tetrahydrate, 98.5% min

Catalog No. AA1222822
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1222822 100 g
AA1222836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1222822 Supplier Thermo Scientific Chemicals Supplier No. 01222822

CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol

Cadmium nitrate is used in manufacturing of cadmium salts, photographic emulsions, laboratory reagent and coloring glass. It is used for porcelain and as a flash powder in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10022-68-1
Molecular Formula CdH2N2O6
Molecular Weight (g/mol) 238.44
MDL Number MFCD00149626
InChI Key IZIPRBSCGMISKX-UHFFFAOYSA-N
Synonym cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
PubChem CID 56924536
ChEBI CHEBI:86156
IUPAC Name cadmium(2+);dinitrate;tetrahydrate
SMILES [Cd++].O[N+]([O-])=O.O[N+]([O-])=O

Specifications

Boiling Point 132°C
Melting Point 59°C
Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 98.5% min.
Solubility Information Highly soluble in water. Soluble in alcohol,acetone,and ammonia.
Formula Weight 308.47 (236.41 Anhydrous)
Odor Weak
Density 2.45
Chemical Name or Material Cadmium nitrate tetrahydrate
Show More Show Less
Hazard Category H301-H312-H330-H340-H350-H360-H372
Hazard Statement GHS H Statement
H301-H350-H272-H312-H332
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P302+P352-P304+P340-P308+P313-P310-P312-P330-P363-P501c
DOTInformation DOT Class: 6.1; Packaging Group: II
EINECSNumber 233-710-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only