missing translation for 'onlineSavingsMsg'
Learn More

Cadmium molybdenum oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Electronic and optical applications

$990.34 - $990.34

Chemical Identifiers

CAS 13972-68-4
Molecular Formula CdMoO4
Molecular Weight (g/mol) 272.36
MDL Number MFCD00015997
InChI Key SQHWLBOPUKZGNS-UHFFFAOYSA-N
Synonym cadmium molybdate, cd.moo4, cadmium 2+ molybdate, cadmium 2+ ;dioxido dioxo molybdenum
PubChem CID 22420716
IUPAC Name cadmium(2+);dioxido(dioxo)molybdenum
SMILES [O--].[O--].[O--].[O--].[Mo].[Cd++]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4020230
View Documents
Thermo Scientific Chemicals
04020230
250 g
Each for $990.34
Only null left
 
Description

Description

Electronic and optical applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13972-68-4
272.36
SQHWLBOPUKZGNS-UHFFFAOYSA-N
22420716
[O--].[O--].[O--].[O--].[Mo].[Cd++]
CdMoO4
MFCD00015997
cadmium molybdate, cd.moo4, cadmium 2+ molybdate, cadmium 2+ ;dioxido dioxo molybdenum
cadmium(2+);dioxido(dioxo)molybdenum

Specifications

∼1250°C
Powder
CdMoO4
UN2570
Slightly soluble in water; Soluble in acid
[O--].[O--].[O--].[O--].[Mo].[Cd++]
272.36
272.35
Odorless
(metals basis)
13972-68-4
250 g
MFCD00015997
cadmium molybdate, cd.moo4, cadmium 2+ molybdate, cadmium 2+ ;dioxido dioxo molybdenum
SQHWLBOPUKZGNS-UHFFFAOYSA-N
cadmium(2+);dioxido(dioxo)molybdenum
22420716
99.5%
Cadmium molybdenum oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c

H302+H312+H332-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS

EINECSNumber : 237-752-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only