missing translation for 'onlineSavingsMsg'
Learn More

Cadmium molybdenum oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog No. AA4020230
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4020230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4020230 Supplier Thermo Scientific Chemicals Supplier No. 04020230
Only null left

Electronic and optical applications

Electronic and optical applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13972-68-4
Molecular Formula CdMoO4
Molecular Weight (g/mol) 272.36
MDL Number MFCD00015997
InChI Key SQHWLBOPUKZGNS-UHFFFAOYSA-N
Synonym cadmium molybdate, cd.moo4, cadmium 2+ molybdate, cadmium 2+ ;dioxido dioxo molybdenum
PubChem CID 22420716
IUPAC Name cadmium(2+);dioxido(dioxo)molybdenum
SMILES [O--].[O--].[O--].[O--].[Mo].[Cd++]

Specifications

Melting Point ∼1250°C
Physical Form Powder
Quantity 250 g
UN Number UN2570
Solubility Information Slightly soluble in water; Soluble in acid
Formula Weight 272.35
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Cadmium molybdenum oxide
Hazard Category H302+H312+H332-H350
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber 237-752-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only