missing translation for 'onlineSavingsMsg'
Learn More

Cadmium iodide, Puratronic™, 99.999% (metals basis)

Catalog No. p-7058903
Change view
Click to view available options
Quantity:
100 g
25 g
Catalog No. Quantity
AA4344214 25 g
AA4344222 100 g
2 options

Catalog No. AA4344214

Supplier: Thermo Scientific Chemicals 04344214

Only null left

CAS: 7790-80-9 | CdI2 | 366.223 g/mol

Cadmium iodide is used in lithography, photography, electroplating and the manufacturing of phosphors, in engraving process, and was used as an antiseptic and astringent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7790-80-9
Molecular Formula CdI2
Molecular Weight (g/mol) 366.223
MDL Number MFCD00010920
InChI Key OKIIEJOIXGHUKX-UHFFFAOYSA-L
Synonym cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25g
PubChem CID 24635
IUPAC Name cadmium(2+);diiodide
SMILES [Cd+2].[I-].[I-]

Specifications

Boiling Point 787°C
Melting Point 387°C to 389°C
Color Brown
Physical Form Powder
Quantity 25 g
UN Number UN2570
Merck Index 14,1621
Solubility Information It is soluble in ethanol,acetone,ether and ammonia.
Formula Weight 366.21
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 5.67 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium iodide
Show More Show Less
Hazard Category H301+H331-H351-H373
Hazard Statement GHS H Statement
H301-H331-H351-H373
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber 232-223-6
RTECSNumber EV1290000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only