Learn More
Gfs Chemicals Inc CADMIUM, METAL, 20-60 MESH, GRANULAR | 100 G | GFS CHEMICALS, INC. Item #: 18591 | Bulk #: 1697
Catalog No. NC9130293
Encompass
CADMIUM, METAL, 20-60 MESH, GRANULAR | Chemical Formula : Cd | CAS # : 7440-43-9 | Specifications : Meets Specs for AWWA/APHA/WEF 4500-NO3-, Particle size 20 - 60 mesh, Shape & app. - granular metallic particles
Catalog No. NC9130293 Supplier Gfs Chemicals Inc Supplier No. 18591
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CADMIUM, METAL, 20-60 MESH, GRANULAR | Chemical Formula : Cd | CAS # : 7440-43-9 | Specifications : Meets Specs for AWWA/APHA/WEF 4500-NO3-, Particle size 20 - 60 mesh, Shape & app. - granular metallic particles