missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride hydrate, 99.99% (metals basis)

Catalog No. p-7049729
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2012914 25 g
AA2012922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2012914 Supplier Thermo Scientific Chemicals Supplier No. 02012914
Only null left

CAS: 654054-66-7 | CdCl2 | 183.31 g/mol

Cadmium chloride hydrate is used in laboratory chemicals and manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 654054-66-7
Molecular Formula CdCl2
Molecular Weight (g/mol) 183.31
MDL Number MFCD00149627
InChI Key YKYOUMDCQGMQQO-UHFFFAOYSA-L
Synonym cadmium chloride, ultra dry, cdcl2.h2o, cadmium chloride hydrate, cadmium chloride 1-hydrate, cadmium chloride hydrate, puratronic, cadmium chloride monohydrate, puriss, cadmium chloride hydrate trace metals basis
PubChem CID 16211517
IUPAC Name dichlorocadmium;hydrate
SMILES [Cl-].[Cl-].[Cd++]

Specifications

Physical Form Powder
Quantity 25 g
UN Number UN2570
Merck Index 14,1617
Solubility Information Soluble in water.
Formula Weight 183.32 (Anhydrous)
Percent Purity 99.99%
Refractive Index 1.6513
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cadmium chloride hydrate
Show More Show Less
Hazard Category H301-H330-H340-H350-H360-H372
Hazard Statement GHS H Statement
H300-H330-H340-H350-H360-H372
Fatal if swallowed.
Fatal if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber 233-296-7
RTECSNumber EV0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only