Learn More
Cadmium chloride hemipentahydrate, ACS reagent
CAS: 7790-78-5 | 2·5 H2O | 228.34 g/mol
$132.30 - $132.30
Chemical Identifiers
| CAS | 7790-78-5 |
|---|---|
| Molecular Formula | 2·5 H2O |
| Molecular Weight (g/mol) | 228.34 |
| MDL Number | MFCD00799071 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC423501000
|
Thermo Scientific Chemicals
423501000 |
100 g | Glass bottle |
Each for $132.30
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7790-78-5 | |
| 2·5 H2O | |
| 228.34 | |
| MFCD00799071 |
Specifications
| 34.0°C | |
| 7790-78-5 | |
| 100 g | |
| CdCl2·5/2H2O | |
| 15, 1622 | |
| 228.34 | |
| 79.5 to 81.0% (CdCl2) | |
| Glass bottle |
| White | |
| Crystals and/or Chunks | |
| 2·5 H2O | |
| MFCD00799071 | |
| Solubility in water: 1680g/L (20°C). Other solubilities: soluble in acetone, slightly soluble in ethanol and methanol | |
| 228.34 | |
| ACS Reagent | |
| Cadmium chloride hemipentahydrate |
Safety and Handling
GHS H Statement
May cause cancer.
May cause genetic defects.
May damage fertility.
May damage the unborn child.
Fatal if inhaled.
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a positi
GHS Signal Word: Danger
RTECSNumber : EV0178000
RUO – Research Use Only