missing translation for 'onlineSavingsMsg'
Learn More

Cadmium chloride hemipentahydrate, ACS reagent

Catalog No. AC423500025
Change view
Click to view available options
Quantity:
100 g
2.5 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423500025 2.5 kg Glass bottle
AC423501000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC423500025 Supplier Thermo Scientific Chemicals Supplier No. 423500025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7790-78-5 | 2·5 H2O | 228.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7790-78-5
Molecular Formula 2·5 H2O
Molecular Weight (g/mol) 228.34
MDL Number MFCD00799071

Specifications

Melting Point 34.0°C
Color White
Physical Form Crystals and/or Chunks
Quantity 2.5 kg
Linear Formula CdCl2·5/2H2O
Merck Index 15, 1622
Solubility Information Solubility in water: 1680g/L (20°C). Other solubilities: soluble in acetone,slightly soluble in ethanol and methanol
Formula Weight 228.34
Percent Purity 79.5 to 81.0% (CdCl2)
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Cadmium chloride hemipentahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
May cause genetic defects.
May damage fertility.
May damage the unborn child.
Fatal if inhaled.
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a positi
RTECSNumber EV0178000

RUO – Research Use Only