missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium chloride hemipentahydrate, ACS, 79.5-81.0%
Description
Cadmium chloride is also used for photocopying, dyeing and electroplating. Useful in pyrotechnic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline |
| Quantity | 50 g |
| Assay Percent Range | 79.5-81.0% |
| Molecular Formula | CdCl2·2.5H2O |
| Solubility Information | Highly soluble in water and slightly soluble in alcohol. |
| Formula Weight | 228.36 (183.32 Anhydrous) |
| Grade | ACS |
| Odor | Odorless |
| Refractive Index | 1.6513 |
| Show More |
Safety and Handling
| Hazard Category | H301-H330-H340-H350-H360FD-H372 |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P304+P340-P308+P313-P310-P330-P501c |
| DOTInformation | DOT Class: 6.1; Packaging Group: I |
| EINECSNumber | 233-296-7 |
| RTECSNumber | EV0175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only