missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Chloride Hemi(Pentahydrate), 79.5 to 81.0%, ACS Reagent, Honeywell™ Fluka™

ACS Reagent, 79.5-81.0%

$809.63 - $809.63

Chemical Identifiers

CAS 7790-78-5
MDL Number MFCD00799071
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60045485
View Documents
Honeywell-Fluka
239208500G
500 g Poly Bottle
Each for $809.63
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7790-78-5
MFCD00799071

Specifications

7790-78-5
CdCl2·21/2H2O
UN2570
79.5 to 81.0%
Poly Bottle
3.327g/cm3; Vapor Density: 6.3 (vs air)
500 g
MFCD00799071
228.36g/mol
ACS Reagent
10mmHg (656°C)
Cadmium Chloride Hemi(Pentahydrate)
Videos
Safety and Handling

Safety and Handling

P201-P260-P273-P284-P301 + P310 + P330-P304 + P340 + P310

H301-H330-H340-H350-H360FD-H372-H410

DOTInformation : Class 6.1 - PG 3 - EHS - Cadmium compounds, HI: all (not BR)

EINECSNumber : 233-296-7

RTECSNumber : EV0178000

SDS
Documents

Documents