missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromide tetrahydrate, Reagent Grade

Catalog No. AA3295814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3295814 25 g
AA3295822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3295814 Supplier Thermo Scientific Chemicals Supplier No. 03295814

CAS: 13464-92-1 | Br2CdH8O4 | 344.282 g/mol

It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13464-92-1
Molecular Formula Br2CdH8O4
Molecular Weight (g/mol) 344.282
MDL Number MFCD00150204
InChI Key PANYFCXNZSEXSU-UHFFFAOYSA-L
Synonym cadmium dibromide, tetrahydrate, acmc-20alji, ksc497c4n, cadmium ii ; bromide tetrahydrate, cadmium 2+ dibromide tetrahydrate, cadmium 2+ ion tetrahydrate dibromide, cadmium bromide, tetrahydrate, reagent
PubChem CID 3084147
IUPAC Name cadmium(2+);dibromide;tetrahydrate
SMILES O.O.O.O.[Br-].[Br-].[Cd+2]

Specifications

Physical Form Powder
Quantity 25 g
UN Number UN2570
Solubility Information Soluble in water.
Formula Weight 344.28 (272.22 Anhydrous)
Grade Reagent
Sensitivity Hygroscopic
Chemical Name or Material Cadmium bromide tetrahydrate
Hazard Category H302+H312+H332-H350
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CADMIUM COMPOUNDS
EINECSNumber 232-165-1
RTECSNumber EU9935000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only