Learn More
Cadmium bromide tetrahydrate, 98%, pure, Thermo Scientific™
$158.08 - $496.40
Chemical Identifiers
| CAS | 13464-92-1 |
|---|---|
| Molecular Formula | Br2Cd·4H2O |
| Molecular Weight (g/mol) | 344.28 |
| InChI Key | PANYFCXNZSEXSU-UHFFFAOYSA-L |
| Synonym | cadmium dibromide, tetrahydrate, acmc-20alji, ksc497c4n, cadmium ii ; bromide tetrahydrate, cadmium 2+ dibromide tetrahydrate, cadmium 2+ ion tetrahydrate dibromide, cadmium bromide, tetrahydrate, reagent |
| PubChem CID | 3084147 |
| IUPAC Name | cadmium(2+);dibromide;tetrahydrate |
| SMILES | O.O.O.O.[Br-].[Br-].[Cd+2] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC453871000
|
Thermo Scientific Chemicals
453871000 |
100g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC453875000
|
Thermo Scientific Chemicals
453875000 |
500g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 13464-92-1 | |
| 344.28 | |
| cadmium dibromide, tetrahydrate, acmc-20alji, ksc497c4n, cadmium ii ; bromide tetrahydrate, cadmium 2+ dibromide tetrahydrate, cadmium 2+ ion tetrahydrate dibromide, cadmium bromide, tetrahydrate, reagent | |
| cadmium(2+);dibromide;tetrahydrate |
| Br2Cd·4H2O | |
| PANYFCXNZSEXSU-UHFFFAOYSA-L | |
| 3084147 | |
| O.O.O.O.[Br-].[Br-].[Cd+2] |
Specifications
| 13464-92-1 | |
| 97.5% min | |
| cadmium dibromide, tetrahydrate, acmc-20alji, ksc497c4n, cadmium ii ; bromide tetrahydrate, cadmium 2+ dibromide tetrahydrate, cadmium 2+ ion tetrahydrate dibromide, cadmium bromide, tetrahydrate, reagent | |
| PANYFCXNZSEXSU-UHFFFAOYSA-L | |
| cadmium(2+);dibromide;tetrahydrate | |
| 3084147 | |
| 98% | |
| Cadmium bromide tetrahydrate |
| 100g | |
| Br2Cd·4H2O | |
| Solubility in water: soluble. | |
| O.O.O.O.[Br-].[Br-].[Cd+2] | |
| 344.28 | |
| 344.28 | |
| Pure |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
GHS Signal Word: Warning
RUO â Research Use Only