missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromide tetrahydrate, 98%, pure

Catalog No. AC453871000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453871000 100 g
AC453875000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453871000 Supplier Thermo Scientific Chemicals Supplier No. 453871000

CAS: 13464-92-1 | Br2Cd·4H2O | 344.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13464-92-1
Molecular Formula Br2Cd·4H2O
Molecular Weight (g/mol) 344.28
InChI Key PANYFCXNZSEXSU-UHFFFAOYSA-L
Synonym cadmium dibromide, tetrahydrate, acmc-20alji, ksc497c4n, cadmium ii ; bromide tetrahydrate, cadmium 2+ dibromide tetrahydrate, cadmium 2+ ion tetrahydrate dibromide, cadmium bromide, tetrahydrate, reagent
PubChem CID 3084147
IUPAC Name cadmium(2+);dibromide;tetrahydrate
SMILES O.O.O.O.[Br-].[Br-].[Cd+2]

Specifications

Quantity 100 g
Assay Percent Range 97.5% min
Solubility Information Solubility in water: soluble.
Formula Weight 344.28
Percent Purity 98%
Grade Pure
Chemical Name or Material Cadmium bromide tetrahydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

RUO – Research Use Only