missing translation for 'onlineSavingsMsg'
Learn More

Cadmium bromide, anhydrous, 98%

Catalog No. AA3296014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3296014 25 g
AA3296022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3296014 Supplier Thermo Scientific Chemicals Supplier No. 03296014

CAS: 7789-42-6 | CdBr2

Cadmium bromide is used in process engraving, lithography and photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-42-6
Molecular Formula CdBr2
MDL Number MFCD00015996
Synonym cadmium 2+ dibromide, cadmium 2+ ion dibromide, cadmium bromide, anhydrous 25g, cadmium bromide, anhydrous,

Specifications

Boiling Point 863°C
Melting Point 567°C
Color White to Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 98.00%
Solubility Information Freely soluble in water,alcohol. Slightly soluble in acetone and ether.
Formula Weight 272.22
Odor Odorless
Sensitivity Hygroscopic
Appearance White to pale yellow powder
Density 5.192 g/cm3
Chemical Name or Material Cadmium bromide
Show More Show Less
Hazard Category H302+H312+H332-H350
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 232-165-1
RTECSNumber EU9935000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only