Learn More
ACROBiosystems Inc Recombinant Protein;Human CADM1 / IGSF4A Protein;HEK293;100UG;CA1,CADM1,CADM-1,BL2,IGSF4,IGSF-4,IGSF4A,NECL2,Necl-2,RA175,ST17,SYNCAM,TSLC1,sTSLC-1,sgIGSF,synCAM1,DKFZp686F1789,MGC149785,MGC518800
Catalog No. 501658995
Encompass
Recombinant Protein;Human CADM1 / IGSF4A Protein;100UG;Human CADM1, His Tag (CA1-H5225) is expressed from human 293 cells (HEK293). It contains AA Gln 45 - His 374 (Accession # NP_055148.3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 38.5 kDa. The protein migrates as 38.5kDa, 50kDa and 55-85 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Catalog No. 501658995 Supplier ACROBiosystems Inc Supplier No. CA1H5225100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Human CADM1 / IGSF4A Protein;100UG;Human CADM1, His Tag (CA1-H5225) is expressed from human 293 cells (HEK293). It contains AA Gln 45 - His 374 (Accession # NP_055148.3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 38.5 kDa. The protein migrates as 38.5kDa, 50kDa and 55-85 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.