Learn More
Cadence Science™ Nonflexible Stainless Steel Animal Feeding Needles
For feeding and dosing without injury to sensitive tissues.
Supplier:  Cadence Science™ POP7924
 View more versions of this product
Catalog No. 14825282
Only null left
Description
Description
Specifications
Chemical Identifiers
Animal Feeding Needle
Reusable
Non-sterile
Stainless Steel (304)
Animal feeding and dosing without injury to sensitive tissues, ideal for pharmacologists, toxicologists, pathologists and other biomedical researchers working with laboratory animals
Specifications
Animal Feeding Needle
Reusable
13G
3 in.
4 mm
Stainless Steel (304)
Animal feeding and dosing without injury to sensitive tissues, ideal for pharmacologists, toxicologists, pathologists and other biomedical researchers working with laboratory animals
Non-sterile
7.62 cm
Product Suggestions
Product Suggestions