Learn More
Cadence Science™ Flexible Plastic (PTFE) Needles with Bite Protectors
Covered at the hub end with 304 stainless steel tubing – protects tubing from damage due to biting during use.
Supplier: Cadence Science™ POP9925B
Chemical Identifiers
| Single-use | |
| Animal feeding | |
| Sterile |
Specifications
| Single-use | |
| 15G | |
| 3.1 in. |
| Animal feeding | |
| Sterile | |
| 2.8 mm |