Learn More
Cadence Science™ Flexible Plastic (PTFE) Needles with Bite Protectors
Covered at the hub end with 304 stainless steel tubing – protects tubing from damage due to biting during use.
Supplier:  Cadence Science™ POP9924B
 View more versions of this product
Catalog No. 14825301
Only null left
Description
Description
Specifications
Chemical Identifiers
Single-use
Animal feeding
Sterile
Specifications
Single-use
18G
3.0 in.
Animal feeding
Sterile
2 mm
Product Suggestions
Product Suggestions