missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadence Science™ Disposable Plastic (PTFE) Animal Feeding Needles
For critical applications requiring sterile, flexible, single-use feeding needles
Supplier: Cadence Science™ POP9920
Description
- Nontoxic, nonpyrogenic
- PTFE tubing is flexible and softer than other types of tubing
- Low friction protects delicate organs
- Translucent for easy confirmation of reagent delivery
- Plastic luer hubs; soft silicone tips
- Sterile, packaged in pouches
- Five needles per pouch, 20 pouches per box
Chemical Identifiers
| Soft, Flexible Animal Feeding Needle | |
| PTFE | |
| Single-use | |
| Sterile |
Specifications
| Soft, Flexible Animal Feeding Needle | |
| Single-use | |
| 20G | |
| 1.5 in. | |
| 0.074 in. |
| PTFE | |
| Animal feeding, For critical applications requiring sterile, flexible, single use feeding needles | |
| Sterile | |
| 0.38 cm | |
| 0.019 cm |