Learn More
Cadence Science™ Deflected Noncoring Septum Penetration Needles
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures.
Supplier:  Cadence Science™ POP4176
 View more versions of this product
Catalog No. 14-825-15N
Only null left
Explore available promotions
Description
Description
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
  • Stainless-steel needles with deflected noncoring septum
  • For noncoring penetration of septa and rubber closures involving air-sensitive and pyrophoric compounds
Specifications
Specifications
Nickel-plated Brass (Hub), Stainless Steel (Tube)
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
6 in.
12 Needles
22 G
Deflected non-coring septum
Penetration Needle
Deflected End
Product Suggestions
Product Suggestions