Learn More
Cadence Science™ Deflected Noncoring Septum Penetration Needles
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures.
Supplier: Cadence Science™ POP4196
Description
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
- Penetration needles with deflected noncoring septum
- For noncoring penetration of septa and rubber closures involving air-sensitive and pyrophoric compounds
Specifications
| Nickel-plated Brass (Hub), Stainless Steel (Tube) | |
| Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures | |
| 12 in. | |
| 12 Needles | |
| Deflected End |
| 18 G | |
| Deflected non-coring septum | |
| Penetration Needle | |
| 30.5 cm |