Learn More
Cadence Science™ Deflected Noncoring Septum Penetration Needles
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures.
Supplier:  Cadence Science™ POP4188
 View more versions of this product
Catalog No. 14-825-15AB
Only null left
Explore available promotions
Description
Description
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
  • Penetration needles with deflected noncoring septum
  • For noncoring penetration of septa and rubber closures involving air-sensitive and pyrophoric compounds
Specifications
Specifications
Nickel-plated Brass (Hub), Stainless Steel (Tube)
Use in gas chromatography and nuclear magnetic resonance spectroscopy procedures
12 in.
12 Needles
Deflected End
20 G
Deflected non-coring septum
Penetration Needle
305 mm
Product Suggestions
Product Suggestions