missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Cacodylic Acid, Sodium Salt, Trihydrate, Calbiochem™,

Catalog No. 205541100GM
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
205541100GM 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 205541100GM Supplier MilliporeSigma™ Supplier No. 205541100GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A carcinogen that also acts as a nephrotoxin. Major metabolite of inorganic arsenics

Chemical Identifiers

CAS 6131-99-3
Molecular Formula C2H12AsNaO5
Molecular Weight (g/mol) 214.024
InChI Key RLGWPHBPRCROJO-UHFFFAOYSA-M
Synonym sodium cacodylate trihydrate, unii-r7a6nc7ygy, cacodylic acid sodium salt trihydrate, r7a6nc7ygy, dimethylarsonic acid sodium salt, cacodylic acid, sodium salt trihydrate, sodium dimethylarsinic acid trihydrate, dimethylarsenic acid sodium salt trihydrate, dimethylarsinic acid sodium salt trihydrate, hydroxydimethylarsine oxide sodium salt trihydrate
PubChem CID 23679059
IUPAC Name sodium;dimethylarsinate;trihydrate
SMILES C[As](=O)(C)[O-].O.O.O.[Na+]

Specifications

Melting Point 195°C to 200°C
Color White
Quantity 100 g
UN Number UN 1688
Solubility Information Easily soluble in cold water
Formula Weight 214
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Sodium cacodylate
Grade Electron microscopy grade
DOTInformation DOT Class: 6.1, Packaging Group: II
EINECSNumber 204-708-2
RTECSNumber CH7700000
Recommended Storage 2° to 8°C (35.6° to 46.4°F); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 4°C (39.2°F).