missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cacodylic acid sodium salt trihydrate 98+%
Description
Cacodylic acid sodium salt is frequently used as a buffering agent during the preparation and fixation of biological samples for transmission in electron microscopy. It is also used as a buffer for formulating screens and for optimization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cacodylic acid sodium salt is frequently used as a buffering agent during the preparation and fixation of biological samples for transmission in electron microscopy. It is also used as a buffer for formulating screens and for optimization.
Solubility
Soluble in water and alcohol. Insoluble in diethyl ether.
Notes
Store in cool place. Hygroscopic and moisture sensitive. Incompatible withstrong oxidizing agents and strong bases.
Cacodylic acid sodium salt is frequently used as a buffering agent during the preparation and fixation of biological samples for transmission in electron microscopy. It is also used as a buffer for formulating screens and for optimization.
Solubility
Soluble in water and alcohol. Insoluble in diethyl ether.
Notes
Store in cool place. Hygroscopic and moisture sensitive. Incompatible withstrong oxidizing agents and strong bases.
Specifications
Specifications
| Linear Formula | (CH3)2AsO2Na·3H2O |
| Quantity | 25 g |
| UN Number | UN1688 |
| Beilstein | 3702348 |
| Merck Index | 14,8595 |
| Solubility Information | Soluble in water and alcohol. Insoluble in diethyl ether. |
| Formula Weight | 214.03 (159.98 Anhydrous) |
| Percent Purity | ≥98% |
| Chemical Name or Material | Cacodylic acid sodium salt trihydrate |
Safety and Handling
| Hazard Category | H301+H331-H350-H373 |
| Hazard Statement | GHS H Statement H301-H331 Toxic if swallowed. Toxic if inhaled. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P281-P301+P310-P308+P313-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SODIUM CACODYLATE |
| EINECSNumber | 204-708-2 |
| RTECSNumber | CH7700000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only