missing translation for 'onlineSavingsMsg'
Learn More

Cacodylic acid sodium salt trihydrate 98+%

Catalog No. AAA1813936
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1813936 500 g
AAA1813914 25 g
AAA1813922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1813936 Supplier Thermo Scientific Chemicals Supplier No. A1813936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 6131-99-3 | C2H12AsNaO5 | 214.024 g/mol

Cacodylic acid sodium salt is frequently used as a buffering agent during the preparation and fixation of biological samples for transmission in electron microscopy. It is also used as a buffer for formulating screens and for optimization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cacodylic acid sodium salt is frequently used as a buffering agent during the preparation and fixation of biological samples for transmission in electron microscopy. It is also used as a buffer for formulating screens and for optimization.

Solubility
Soluble in water and alcohol. Insoluble in diethyl ether.

Notes
Store in cool place. Hygroscopic and moisture sensitive. Incompatible withstrong oxidizing agents and strong bases.

Chemical Identifiers

CAS 6131-99-3
Molecular Formula C2H12AsNaO5
Molecular Weight (g/mol) 214.024
MDL Number MFCD00149079
InChI Key RLGWPHBPRCROJO-UHFFFAOYSA-M
Synonym sodium cacodylate trihydrate, unii-r7a6nc7ygy, cacodylic acid sodium salt trihydrate, r7a6nc7ygy, dimethylarsonic acid sodium salt, cacodylic acid, sodium salt trihydrate, sodium dimethylarsinic acid trihydrate, dimethylarsenic acid sodium salt trihydrate, dimethylarsinic acid sodium salt trihydrate, hydroxydimethylarsine oxide sodium salt trihydrate
PubChem CID 23679059
IUPAC Name sodium;dimethylarsinate;trihydrate
SMILES C[As](=O)(C)[O-].O.O.O.[Na+]

Specifications

Linear Formula (CH3)2AsO2Na·3H2O
Quantity 500 g
UN Number UN1688
Beilstein 3702348
Merck Index 14,8595
Solubility Information Soluble in water and alcohol. Insoluble in diethyl ether.
Formula Weight 214.03 (159.98 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Cacodylic acid sodium salt trihydrate
Hazard Category H301+H331-H350-H373
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SODIUM CACODYLATE
EINECSNumber 204-708-2
RTECSNumber CH7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only