missing translation for 'onlineSavingsMsg'
Learn More

Cacodylic acid, 98%

Catalog No. AC318150500
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC318150500 50 g Glass bottle
AC318152500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC318150500 Supplier Thermo Scientific Chemicals Supplier No. 318150500

CAS: 75-60-5 | C2H7AsO2 | 138.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-60-5
Molecular Formula C2H7AsO2
Molecular Weight (g/mol) 138.00
MDL Number MFCD00002095
InChI Key OGGXGZAMXPVRFZ-UHFFFAOYSA-N
Synonym cacodylic acid, hydroxydimethylarsine oxide, chexmate, silvisar, phytar, arsan, dimethylarsenic acid, erase, arsinic acid, dimethyl, silvisar 510
PubChem CID 2513
ChEBI CHEBI:48765
IUPAC Name dimethylarsinic acid
SMILES C[As](C)(O)=O

Specifications

Melting Point 195°C to 198°C
Color White
Assay Percent Range 98%
Linear Formula (CH3)2As(O)OH
Merck Index 15, 1609
Formula Weight 138.01
Percent Purity ≥97.5%
Quantity 50 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Chemical Name or Material Cacodylic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only