Learn More
Cacodylic acid, 98%
CAS: 75-60-5 | C2H7AsO2 | 138.00 g/mol
$145.52 - $480.38
Chemical Identifiers
| CAS | 75-60-5 |
|---|---|
| Molecular Formula | C2H7AsO2 |
| Molecular Weight (g/mol) | 138.00 |
| MDL Number | MFCD00002095 |
| InChI Key | OGGXGZAMXPVRFZ-UHFFFAOYSA-N |
| Synonym | cacodylic acid, hydroxydimethylarsine oxide, chexmate, silvisar, phytar, arsan, dimethylarsenic acid, erase, arsinic acid, dimethyl, silvisar 510 |
| PubChem CID | 2513 |
| ChEBI | CHEBI:48765 |
| IUPAC Name | dimethylarsinic acid |
| SMILES | C[As](C)(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1207588
|
Thermo Scientific Chemicals
A1207588 |
2.5 g |
N/A
|
|
|||||
|
AAA1207509
|
Thermo Scientific Chemicals
A1207509 |
10 g |
N/A
|
|
|||||
Description
A useful arsenic acid for proteomics research. It a good substitute for phosphate in applications to avoid phosphates. Cacodylic Acid is useful for some DNA applications and is also popular in microscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA useful arsenic acid for proteomics research. It a good substitute for phosphate in applications to avoid phosphates. Cacodylic Acid is useful for some DNA applications and is also popular in microscopy.
Solubility
Soluble in water (50 mg/ml), water (2000 at 25°C), alcohols, and acetic acid. Insoluble in ether.
Notes
Hygroscopic. Store away from moisture, air and oxidizing agents. Store under inert gas.
Chemical Identifiers
| 75-60-5 | |
| 138.00 | |
| OGGXGZAMXPVRFZ-UHFFFAOYSA-N | |
| 2513 | |
| dimethylarsinic acid |
| C2H7AsO2 | |
| MFCD00002095 | |
| cacodylic acid, hydroxydimethylarsine oxide, chexmate, silvisar, phytar, arsan, dimethylarsenic acid, erase, arsinic acid, dimethyl, silvisar 510 | |
| CHEBI:48765 | |
| C[As](C)(O)=O |
Specifications
| 200°C | |
| C2H7AsO2 | |
| UN1572 | |
| 14,1604 | |
| Soluble in water (50mg/ml),water (2000 at 25°C),alcohols,and acetic acid. Insoluble in ether. | |
| C[As](C)(O)=O | |
| 138.00 | |
| CHEBI:48765 | |
| 98% | |
| Hygroscopic |
| 75-60-5 | |
| MFCD00002095 | |
| 1736965 | |
| cacodylic acid, hydroxydimethylarsine oxide, chexmate, silvisar, phytar, arsan, dimethylarsenic acid, erase, arsinic acid, dimethyl, silvisar 510 | |
| OGGXGZAMXPVRFZ-UHFFFAOYSA-N | |
| dimethylarsinic acid | |
| 2513 | |
| 138 | |
| 2.5 g | |
| Cacodylic acid |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CACODYLIC ACID
EINECSNumber : 200-883-4
RTECSNumber : CH7525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only