missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cacodylic acid, 98%
Description
A useful arsenic acid for proteomics research. It a good substitute for phosphate in applications to avoid phosphates. Cacodylic Acid is useful for some DNA applications and is also popular in microscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A useful arsenic acid for proteomics research. It a good substitute for phosphate in applications to avoid phosphates. Cacodylic Acid is useful for some DNA applications and is also popular in microscopy.
Solubility
Soluble in water (50 mg/ml), water (2000 at 25°C), alcohols, and acetic acid. Insoluble in ether.
Notes
Hygroscopic. Store away from moisture, air and oxidizing agents. Store under inert gas.
A useful arsenic acid for proteomics research. It a good substitute for phosphate in applications to avoid phosphates. Cacodylic Acid is useful for some DNA applications and is also popular in microscopy.
Solubility
Soluble in water (50 mg/ml), water (2000 at 25°C), alcohols, and acetic acid. Insoluble in ether.
Notes
Hygroscopic. Store away from moisture, air and oxidizing agents. Store under inert gas.
Specifications
Specifications
| Melting Point | 200°C |
| UN Number | UN1572 |
| Beilstein | 1736965 |
| Merck Index | 14,1604 |
| Solubility Information | Soluble in water (50mg/ml),water (2000 at 25°C),alcohols,and acetic acid. Insoluble in ether. |
| Formula Weight | 138 |
| Percent Purity | 98% |
| Quantity | 10 g |
| Sensitivity | Hygroscopic |
| Chemical Name or Material | Cacodylic acid |
Safety and Handling
| Hazard Category | H301+H331-H350 |
| Hazard Statement | GHS H Statement H301-H331 Toxic if swallowed. Toxic if inhaled. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CACODYLIC ACID |
| EINECSNumber | 200-883-4 |
| RTECSNumber | CH7525000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only